Kimcote Culture Tubes
  • Class A glass gives uniform thickness
Ultra-clear safety-coated glassware helps stop and control spills in the event of glassware breakage. If a break does occur, the durable odor-free glassware will reduce the hazards of shattered glass and leakage of toxic or corrosive chemicals. Glassware is ideal for chemical storage, transportation, and handling of hazardous chemicals, harmful biologicals, and acids. Glassware has a unique nonslip surface when wet or dry.Manufactured from KG-33 borosilicate glass for maximum heat transfer and superior chemical resistance. Meets ASTM E982, Type VI requirements. Rubber-lined screw caps are included and are unattached. Autoclavable to120°C (250°F).
117,000.00 - ₹211,000.00INR / Case of 144
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Tube Bottom Shape Tube Capacity (mL) Tube Material
Availability Pricing
DWK Life Sciences (Kimble) KC45066-13100 screw thread, culture tube with unattached rubber-lines caps, 8 mL 3457230 Mfr # KC45066-13100
COMPARE
Round 8 Glass
₹211,000.00INR / Case of 144
DWK Life Sciences (Kimble) KC45066-16125 Culture Tube, plastic-coated glass, 16 mL, 144/cs 3457232 Mfr # KC45066-16125
COMPARE
Round 16 Glass
₹209,000.00INR / Case of 144
DWK Life Sciences (Kimble) KC45066-25150 Culture Tube, plastic-coated glass, 50 mL, 48/cs 3457236 Mfr # KC45066-25150
COMPARE
Round 50 Glass
₹117,000.00INR / Case of 48