Kimcote Safety Coated Erlenmeyer Flasks
KimCote™ plastic coating provides added safety
Ultra-clear safety-coated flasks helps stop and control spills in the event of glassware breakage. If a break does occur, the durable odor-free glassware will reduce the hazards of shattered glass and leakage of toxic or corrosive chemicals. Glassware is ideal for chemical storage, transportation, and handling of hazardous chemicals, harmful biologicals, and acids.
Glassware has a unique nonslip surface when wet or dry. Durable safety coated flasks feature large opening and GL 45 threaded closures for easy pouring and tight sealing. Graduations are in durable white enamel. Blue caps and clear pour rings are included. Autoclavable to120°C (250°F).
28,200.00 - ₹54,600.00INR / Case of 6
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Graduation Interval (mL) Material
Availability Pricing
DWK Life Sciences (Kimble) KC26720-250 Erlenmeyer Flask with Screw Caps, 250 mL 3452441 Mfr # KC26720-250
₹38,900.00INR / Case of 6
COMPARE
250 25 Glass
Call 91-22-61394444 for Availability
₹38,900.00INR / Case of 6
DWK Life Sciences (Kimble) KC26720-500 Erlenmeyer Flask with Screw Caps, 500 mL 3452443 Mfr # KC26720-500
₹45,100.00INR / Case of 6
COMPARE
500 50 Glass
Call 91-22-61394444 for Availability
₹45,100.00INR / Case of 6
DWK Life Sciences (Kimble) KC26720-1000 Erlenmeyer Flask with Screw Caps, 1000 mL 3452445 Mfr # KC26720-1000
₹54,600.00INR / Case of 4
COMPARE
1000 50 Glass
Call 91-22-61394444 for Availability
₹54,600.00INR / Case of 4
DWK Life Sciences (Kimble) KC26720-2000 Erlenmeyer Flask with Screw Caps, 2000 mL 3452446 Mfr # KC26720-2000
₹28,200.00INR / Case of 2
COMPARE
2000 100 Glass
Call 91-22-61394444 for Availability
₹28,200.00INR / Case of 2