Kinesis® Short Thread Polypropylene Vials, 9 mm
Compatible with all autosamplers
  • Excellent alternative to glass vials
Short thread vials have a wide opening (6.0 mm ID) with a unique thread design that allows you to screw on the caps and still use them in a robotic autosampler. The thread sits much higher on the vial allowing space between the bottom of the thread and the shoulder of the vial. PP vials are ideal for samples where glass is not appropriate; as with samples that adhere to glass.
21,100.00 - ₹42,900.00INR / Pkg of 1000
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Tube Bottom Shape Tube Color Volume (mL)
Availability Pricing
COMPARE
Slightly concave Clear 2
₹37,100.00INR / Pkg of 1000
COMPARE
V-shape Clear 0.3
₹21,100.00INR / Pkg of 1000
COMPARE
V-shape Amber 0.3
₹42,900.00INR / Pkg of 1000
COMPARE
Flat Clear 0.7
₹35,400.00INR / Pkg of 1000