Cole-Parmer® TELOS® QuEChERS Extraction Kits
Quickly and economically extract the required analytes into solution
  • Salt pack helps transfer salt to the centrifuge tube quickly and easily with minimal waste
  • Conform with documented AOAC 2007.01 and EN 15662 protocols
  • Break up complex sample types with homogenizer balls
TELOS QuEChERS extraction kits offer the first stage cleanup for many sample types. This initial crude extraction and partitioning step uses organic solvent and salt solution to extract the required analytes into solution. Select from extraction kits without a tube, kits with a 50 mL centrifuge tube, or kits with a tube and homogenizer beads. Beads aid in breaking up more complex sample types.
17,200.00 - ₹36,200.00INR / Pkg of 50
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Salts Methodology Description
Availability Pricing
COMPARE
6 g MgSO4 (anhydrous), 1.5 g NaAc.3H2O AOAC 2007.01 Kit without tubes
₹17,200.00INR / Pkg of 50
COMPARE
6 g MgSO4 (anhydrous), 1.5 g NaAc (anhydrous) AOAC 2007.01 Kit with 50 mL centrifuge tubes
₹17,600.00INR / Pkg of 50
COMPARE
6 g MgSO4 (anhydrous), 1.5 g NaAc (anhydrous) AOAC 2007.01 Kit without tubes
₹29,600.00INR / Pkg of 50
COMPARE
4 g MgSO4 (anhydrous), 1 g NaCl, 0.5 g disodium citrate sesquihydrate (TSCD), 1 g sodium citrate (DHS) EN 15662 Kit with 50 mL centrifuge tubes
₹36,200.00INR / Pkg of 50
COMPARE
4 g MgSO4 (anhydrous), 1 g NaCl, 0.5 g disodium citrate sesquihydrate (TSCD), 1 g sodium citrate (DHS) EN 15662 Kit without tubes
₹30,300.00INR / Pkg of 50
COMPARE
4 g MgSO4 (anhydrous), 1 g NaCl, 0.5 g disodium citrate sesquihydrate (TSCD), 1 g sodium citrate (DHS) EN 15662 Kit with 50 mL centrifuge tubes and homogenizer beads
₹33,900.00INR / Pkg of 50