Micro Bunsen Burners with Adjustable Orifice
Microburners include adjustable air supply and tapered serrated gas connection. Optional tripod stand 36310-50 supports a 1-L beaker.
7,000.00 - ₹7,300.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
BTU Output (per hour) Gas Type Inlet Tube OD (in)
Availability Pricing
Humboldt H-5810 Accuflame Bunsen Burner, Micro-Burner, Natural Gas 3613030 Mfr # H-5810
COMPARE
1200 Natural Gas 0.3125
₹7,000.00INR / Each
Humboldt H-5815 Accuflame Micro Bunsen Burner with Adjustable Orifice; Lp Gas 3613032 Mfr # H-5815
COMPARE
3200 Liquid Propane (LP) 0.3125
₹7,300.00INR / Each