Microtorch Kit
Hand-held or table-top butane torch
  • Meets all consumer products safety commission (CPSC) requirements
  • Adjustable broad or pinpoint flame
This refillable microtorch produces an adjustable wind resistant flame with temperatures up to 2500°F (1300°C). The pinpoint flame is perfect for heat shrinking and terminating as well as general heating applications. The torch features a built-in self-igniting system with gas shut-off safety lock, a extra wide base perfect for hands-free use and a durable fuel tank containing enough fuel for up to 2 hours of usage. Torch measure 6.1" H x 3" W x 1.3" D
Call For Price
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Gas Type Description
Availability Pricing
Master Appliance MT-51B Microtorch with Butane Fuel 0303051 Mfr # MT-51B
COMPARE
Butane Microtorch with Butane Fuel
Call For Price
Master Appliance MT-76K Trigger Torch Heat Kit 0303053 Mfr # MT-76K
COMPARE
Butane Trigger Torch Heat Kit