Millipore Amicon Ultra Centrifugal Filters
Low-binding regenerated cellulose membrane, fast processing, and >90% recoveries
  • Unique vertical housing
These units are premier tools for protein concentration and desalting. Typical applications include purifying macromolecular components from tissue culture extracts or cell lysates and concentrating biological samples.
61,300.00 - ₹121,000.00INR / Each
19 variations of this product are available.
See All Product Options Below
19 PRODUCT OPTIONS
Compareitem specifications
Nominal Molecular Weight Limit (Mwl) (Dalton) Tube Size (mL)
Availability Pricing
EMD Millipore UFC500324 Amicon Ultra Centrifugal Filters, 0.5 mL, 3K NMWL; pack of 24 2996820 Mfr # UFC500324
COMPARE
3000 0.5
₹75,100.00INR / Pkg of 24
EMD Millipore UFC501024 Amicon Ultra Centrifugal Filters, 0.5 mL, 10K MWCO; pack of 24 2996822 Mfr # UFC501024
COMPARE
10000 0.5
₹72,400.00INR / Pkg of 24
EMD Millipore UFC503024 Amicon Ultra Centrifugal Filters, 0.5 mL, 30K MWCO; pack of 24 2996824 Mfr # UFC503024
COMPARE
30000 0.5
₹77,100.00INR / Pkg of 24
EMD Millipore UFC505024 Amicon Ultra Centrifugal Filters, 0.5 mL, 50K MWCO; pack of 24 2996826 Mfr # UFC505024
COMPARE
50000 0.5
₹77,100.00INR / Pkg of 24
EMD Millipore UFC510024 Amicon Ultra Centrifugal Filters, 0.5 mL, 100K MWCO; pack of 24 2996828 Mfr # UFC510024
COMPARE
100000 0.5
₹90,500.00INR / Pkg of 24
EMD Millipore UFC200324 Amicon Ultra Centrifugal Filters, 2.0 mL, 3K MWCO; pack of 24 2996830 Mfr # UFC200324
COMPARE
3000 2
₹85,000.00INR / Pkg of 24
EMD Millipore UFC203024 Amicon Ultra Centrifugal Filters, 2.0 mL, 30K MWCO; pack of 24 2996834 Mfr # UFC203024
COMPARE
30000 2
₹85,000.00INR / Pkg of 24
EMD Millipore UFC205024 Amicon Ultra Centrifugal Filters, 2.0 mL, 50K MWCO; pack of 24 2996836 Mfr # UFC205024
COMPARE
50000 2
₹78,100.00INR / Pkg of 24
EMD Millipore UFC210024 Amicon Ultra Centrifugal Filters, 2.0 mL, 100K MWCO; pack of 24 2996838 Mfr # UFC210024
COMPARE
100000 2
₹88,500.00INR / Pkg of 24
EMD Millipore UFC801024 Amicon Ultra Centrifugal Filters, 4 mL, 10K MWCO; pack of 24 2996972 Mfr # UFC801024
COMPARE
10000 4
₹61,300.00INR / Each
EMD Millipore UFC803024 Amicon Ultra Centrifugal Filters, 4 mL, 30K MWCO; pack of 24 2996974 Mfr # UFC803024
COMPARE
30000 4
₹72,500.00INR / Each
EMD Millipore UFC805024 Amicon Ultra Centrifugal Filters, 4 mL, 50K MWCO; pack of 24 2996976 Mfr # UFC805024
COMPARE
50000 4
₹66,800.00INR / Each
EMD Millipore UFC810024 Amicon Ultra Centrifugal Filters, 4 mL, 100K MWCO; pack of 24 2996978 Mfr # UFC810024
COMPARE
100000 4
₹72,500.00INR / Each
EMD Millipore UFC901024 Amicon Ultra Centrifugal Filters, 15 mL, 10K MWCO; pack of 24 2996982 Mfr # UFC901024
COMPARE
10000 15
₹113,000.00INR / Each
EMD Millipore UFC903024 Amicon Ultra Centrifugal Filters, 15 mL, 30K MWCO; pack of 24 2996984 Mfr # UFC903024
COMPARE
30000 15
₹121,000.00INR / Each
EMD Millipore UFC905024 Amicon Ultra Centrifugal Filters, 15 mL, 50K MWCO; pack of 24 2996986 Mfr # UFC905024
COMPARE
50000 15
₹121,000.00INR / Each
EMD Millipore UFC910024 Amicon Ultra Centrifugal Filters, 15 mL, 100K MWCO; pack of 24 2996988 Mfr # UFC910024
COMPARE
100000 15
₹113,000.00INR / Each
EMD Millipore UFC800324 Amicon Ultra Centrifugal Filters, 4 mL, 3K NMWL; pack of 24 2996990 Mfr # UFC800324
COMPARE
3000 4
₹66,800.00INR / Pkg of 24
EMD Millipore UFC900324 Amicon Ultra Centrifugal Filters, 15 mL, 3K NMWL; pack of 24 2996992 Mfr # UFC900324
COMPARE
3000 15
₹107,000.00INR / Pkg of 24
Showing 1- 19 of 19