Millipore® Ultrafiltration Disc Membranes
Ideal for stirred cells and micropartition system
  • Tight microstructure ensures the highest possible retention with the lowest possible adsorption of protein, DNA, or other macromolecules
These high-recovery hydrophilic membranes are made of regenerated cellulose. Use when concentrating or desalting extremely dilute solutions or whenever your sample is hydrophobic.
Call For Price
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Diameter (mm) Nominal Molecular Weight Limit (Mwl) (Dalton)
Availability Pricing
EMD Millipore PLAC02510 Ultrafiltration Disc Membranes, 1k 25mm 10pk 2994900 Mfr # PLAC02510
Call For Price
COMPARE
25 1000
Loading
Call For Price
EMD Millipore PLBC04310 Ultrafiltration Disc Membranes, 3k 43mm 10pk 2994912 Mfr # PLBC04310
Call For Price
COMPARE
44.5 3000
Loading
Call For Price
EMD Millipore PLBC06210 Ultrafiltration Disc Membranes, 3k 10pk 2994914 Mfr # PLBC06210
Call For Price
COMPARE
63.5 3000
Loading
Call For Price
EMD Millipore PLTK02510 Ultrafiltration Disc Membranes, 30k 25mm 10pk 2994924 Mfr # PLTK02510
Call For Price
COMPARE
25 30000
Loading
Call For Price
EMD Millipore PLTK04310 Ultrafiltration Disc Membranes, 30k 43mm 10pk 2994926 Mfr # PLTK04310
Call For Price
COMPARE
44.5 30000
Loading