Minntech Hollow Fiber Filter Capsules and Cartridges
Provide high flow rates with low pressure drop
  • Steam sterilize capsules in-line or autoclave off-line
  • Polysulfone membrane offers low protein binding
These new hollow fiber filters offer superior performance in a compact size. Filters have up to six times more surface area compared to other pleated membrane filters, providing greater capacity and longer filter life. The unique membrane structure helps remove bacteria, high-level endotoxins, pyrogens, and spores — ideal for a variety of water purification applications. Maximum differential pressure is 30 psi (2 bar). Capsules and cartridges provided nonsterile.
150,000.00 - ₹833,000.00INR / Case of 6
14 variations of this product are available.
See All Product Options Below
14 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Connections Pore Size (micron)
Availability Pricing
Minntech 279-30-009 Hollow Fiber Filter Capsule, 0.5-2.5L, 0.1um, 1/4" NPTM; 6/Pk 2988008 Mfr # 279-30-009
COMPARE
1/4" NPT(M) 0.1
₹678,000.00INR / Case of 6
Minntech 279-30-111 Hollow Fiber Filter Capsule, 0.5-2.5L, 0.2um, 3/8" ID; 6/Pk 2988010 Mfr # 279-30-111
COMPARE
3/8" hose barb 0.2
₹550,000.00INR / Case of 6
Minntech 279-30-007 Hollow Fiber Filter Capsule, 0.5-2.5L, 0.05um, 1/4" NPTM; 6/Pk 2988012 Mfr # 279-30-007
COMPARE
1/4" NPT(M) 0.05
₹712,000.00INR / Case of 6
Minntech 279-30-011 Hollow Fiber Filter Capsule, 0.5-2.5L, 0.2um, 1/4"NPTM; 6/Pk 2988022 Mfr # 279-30-011
COMPARE
1/4" NPT(M) 0.2
₹623,000.00INR / Case of 6
Minntech 279-20-029 Hollow Fiber Cartridge Filter, 0.20 µm Pore Size 2988025 Mfr # 279-20-029
COMPARE
replacement filter only 0.2
₹150,000.00INR / Each
Minntech 279-20-001 Hollow Fiber Cartridge Filter, 0.05 µm Pore Size 2988026 Mfr # 279-20-001
COMPARE
replacement filter only 0.05
₹280,000.00INR / Each
Minntech 279-30-001 Hollow Fiber Filter Capsule, 1-5.7L, 0.05um, 1/4"NPTM; 6/Pk 2988028 Mfr # 279-30-001
COMPARE
1/4" NPT(M) 0.05
₹748,000.00INR / Case of 6
Minntech 279-30-101 Hollow Fiber Filter Capsule, 1-5.7L, 0.05um, 3/8"NPTM; 6/Pk 2988030 Mfr # 279-30-101
COMPARE
3/8" NPT(M) 0.05
₹748,000.00INR / Case of 6
Minntech 279-30-003 Hollow Fiber Filter Capsule, 1-5.7L, 0.1um, 1/4"NPTM; 6/Pk 2988032 Mfr # 279-30-003
COMPARE
1/4" NPT(M) 0.1
₹817,000.00INR / Case of 6
Minntech 279-30-103 Hollow Fiber Filter Capsule, 1-5.7L, 0.1um, 3/8"NPTM; 6/Pk 2988034 Mfr # 279-30-103
COMPARE
3/8" NPT(M) 0.1
₹833,000.00INR / Case of 6
Minntech 279-30-005 Hollow Fiber Filter Capsule, 1-5.7L, 0.2um, 1/4"NPTM; 6/Pk 2988036 Mfr # 279-30-005
COMPARE
1/4" NPT(M) 0.2
₹717,000.00INR / Case of 6
Minntech 279-30-105 Hollow Fiber Filter Capsule, 1-5.7L, 0.2um, 3/8"NPTM; 6/Pk 2988038 Mfr # 279-30-105
COMPARE
3/8" NPT(M) 0.2
₹753,000.00INR / Case of 6
Minntech 279-20-005 Hollow fiber cartridge filter; 0.05 µm, 20"L 2998002 Mfr # 279-20-005
COMPARE
replacement filter only 0.05
₹340,000.00INR / Each
Minntech 279-20-033 Hollow fiber cartridge filter; 0.2 µm, 20"L 2998010 Mfr # 279-20-033
COMPARE
replacement filter only 0.2
₹173,000.00INR / Each
Showing 1- 14 of 14