Motic® Microscope C-Mount Adaptors
Easily connect your trinocular microscope to a camera
  • Compatible with Motic® trinocular microscopes
In order to use your microscope camera with the trinocular port on your microscope, you need a C-mount adapter to join the microscope and camera together. The C-mount adapter that is needed will depend on your microscope, as well as your camera.
38,500.00 - ₹63,400.00INR / Each
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Compatible With Description
Availability Pricing
Motic Instruments Microscope Camera Adaptor, C-Mount, 0.65X, for 2/3" Chip Sensor 4841203 Mfr # 1101001901781
₹41,700.00INR / Each
COMPARE
Motic BA Series, AE31E/AE2000, and SMZ-161/171 trinocular microscopes Microscope Camera Adaptor, C-Mount, 0.65X, for 2/3" Chip Sensor
Loading
₹41,700.00INR / Each
Motic Instruments Microscope Camera Adaptor, C-Mount, 0.5X, for 1/3" and 1/2" Chip Sensor 4841206 Mfr # 1101001901791
₹43,700.00INR / Each
COMPARE
Motic BA Series, AE31E/AE2000, and SMZ-161/171 trinocular microscopes Microscope Camera Adaptor, C-Mount, 0.5X, for 1/3" and 1/2" Chip Sensor
Loading
₹43,700.00INR / Each
Motic Instruments Microscope Camera Adaptor, C-Mount, 0.35X, for 1/3” Chip Sensors 4892590 Mfr # 1101001904115
₹63,400.00INR / Each
COMPARE
Motic BA Series, AE31E/AE2000, and SMZ-161/171 trinocular microscopes Microscope Camera Adaptor, C-Mount, 0.35X, for 1/3” Chip Sensors
Loading
₹63,400.00INR / Each
Motic Instruments Microscope Camera Adaptor, C-Mount, 0.35X, for 1/3” Chip Sensors 4892591 Mfr # 1101010100861
₹38,500.00INR / Each
COMPARE
Motic SMZ160-BP, SMZ160-BLED, SMZ160-TP, and SMZ160-TLED trinocular microscopes Microscope Camera Adaptor, C-Mount, 0.35X, for 1/3” Chip Sensors
Loading
₹38,500.00INR / Each
Motic Instruments Microscope Camera Adaptor, C-Mount, 0.5X, for 1/3" and 1/2" Chip Sensor 4892592 Mfr # 1101010100871
₹38,500.00INR / Each
COMPARE
Motic SMZ160-BP, SMZ160-BLED, SMZ160-TP, and SMZ160-TLED trinocular microscopes Microscope Camera Adaptor, C-Mount, 0.5X, for 1/3" and 1/2" Chip Sensor
Loading
₹38,500.00INR / Each