Motic® Moticam Microscope Camera with Tablet
Capture, view, edit, and share live images with Wi-Fi enabled camera and Android™ tablet
  • Documentation and analysis for a deeper view of your slide
  • Simply log on with your tablet and begin to capture, edit, and save your images
  • Perfect for the immediate presentation of image results
Motic® Moticam microscope Wi-Fi camera with Android™ tablet is designed to display images and video directly on the tablet or transfer to Wi-Fi enabled devices. This setup is perfect for the immediate presentation of image results to colleagues. Detaching the tablet for an even more flexible application is possible thanks to the Wi-Fi data transfer. Use the pre-installed MotiConnect App and get ready to capture, edit and share your microscope images from the tablet in a few taps. These cameras can produce still image and video capture, live and still image measurement, image adjustments, white balance, automatic & manual exposure, individual objective calibration system.
176,000.00 - ₹246,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Data Transfer Image Sensor Size
Availability Pricing
Motic Instruments Moticam BTX5-8 Detachable Tablet Camera System for Trinocular Microscopes, Wi-Fi, 8" LCD, 1280 x 800 4892620 Mfr # D-MOTICAM BTX5-8
COMPARE
Wi-Fi (2.4 & 5 GHz), Ethernet 1/3" CMOS 8" 1280 x 800 Tablet
₹176,000.00INR / Each
Motic Instruments Moticam BTX5-10 Detachable Tablet Camera System for Trinocular Microscopes, Wi-Fi, 10" LCD, 1280 x 800 4892621 Mfr # D-MOTICAM BTX5-10
COMPARE
Wi-Fi (2.4 & 5 GHz), Ethernet 1/3" CMOS 10" 1280 x 800 Tablet
₹246,000.00INR / Each