MTC Bio Handheld Laboratory Label Printers
Print labels on the spot for quick identification
  • Built-in cutter
  • Lightweight, handheld operation
  • Handles a variety of label cassettes for various tube sizes
  • Able to print a variety of barcodes
The handheld lab printer is ideal for labeling cryo tubes, tissue cassettes, slides, and flasks. Just type in the desired copy using the 50 plus button keypad and press print. Within seconds your label is printed and is ready for application. A built-in cutter allows you to easily cut the label from the tape. The printer is powered by six AA batteries or a 9V power supply.
Choose from 13 label cassettes (sold separately) available in a variety of tape colors, including transparent. Three label heights are available for different sized vessels. The 6 mm tapes are great for tissue culture plates, 12 mm tapes are ideal from microtubes, and the extra tall 24 mm tapes are ideal for large tubes and flasks. Special "ultra-low" label tapes are also available for storage in -80 °C environments, or for liquid nitrogen applications.
49,600.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Power (VAC) Description
Availability Pricing
MTC Bio LABeler™ Handheld Laboratory Printer w/Power Supply, US Plug 0653051 Mfr # L9010
COMPARE
120 Handheld Laboratory Printer w/Power Supply, US Plug
₹49,600.00INR / Each
MTC Bio LABeler™ Handheld Laboratory Printer w/Power Supply, EU Plug 0653052 Mfr # L9010E
COMPARE
220 Handheld Laboratory Printer w/Power Supply, EU Plug
₹49,600.00INR / Each