Multifunction Temperature Dataloggers
Over/under alarm threshold settings provide added versatility
  • PC software features stop on full, record continuously, and memory rollover modes
  • Compact design fits almost anywhere
These dataloggers are ideal for monitoring conditions in laboratories, pharmaceutical storage and clean rooms, refrigerators, freezers, museums, manufacturing environments and more. Conduct real time data monitoring with required Windows™ based PC software and interface (order separately below) or export data to text or excel for further analysis. Windows™based software is compatible with Windows™ 95 and above and is required to program and run all dataloggers. Minimum system requirements include 8 MB RAM, 2 MB disk space, 800 x 600 resolution, and COM or USB port. Includes CD ROM, LINT interface, and 6-ft cable. All loggers feature custom user identification, visual alarm indication, over/under alarm threshold settings, user selectable recording start, data rollover, unique logger ID, zoom capability, and user-selectable sampling rates.
60,400.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Temperature (° F) Temperature Accuracy (° F) Temperature Resolution (° F)
Availability Pricing
Supco LT2 Temperature Data Logger with Alarm 2303750 Mfr # LT2
COMPARE
150 ±1.0 0.1
Call For Price
Supco LTT Dual-Channel Temperature Data Logger with Alarm 2303752 Mfr # LTT
COMPARE
300 ±2 0.1
Call For Price
Supco LPT Temperature/Pressure Data Logger with Alarm 2303758 Mfr # LPT
COMPARE
150 ±1 0.1
₹60,400.00INR / Each