Neptune Drum and Barrel Heavy-Duty Air-Drive Motor Mixers
Air-drive motor Mixers generate up to 1/2 hp at 80 psi (maximum pressure), 1/3 hp at 60 psi, and 1/5 hp at 40 psi. We recommend using an air filter/regulator and lubricator—order 07042-75 and 07042-70 separately.
200,000.00 - ₹287,000.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Impeller Size (in) Mounting Type Shaft Length (in)
Availability Pricing
Neptune Drum/barrel heavy-duty, air-drive mixer with drum lip mounting 0431840 Mfr # F-4.0
COMPARE
3.75 Drum-lip clamp 33.125
₹238,000.00INR / Each
Neptune Drum/barrel heavy-duty, air-drive mixer with C-clamp 0431850 Mfr # B-4.0
COMPARE
4 C-clamp 35
₹200,000.00INR / Each
Neptune Drum/barrel heavy-duty, air-drive mixer with 2" NPT(M) mounting 5050020 Mfr # E-4.0
COMPARE
3.75 2" NPT(M) 29
₹287,000.00INR / Each