Neptune Heavy-Duty, Gear-Drive Air-Powered Mixers
Powerful mixers for difficult applications
  • Grease-filled gearbox eliminates oil seal and the possibility of oil contamination
  • Mixers have no clutches to slip, wear, or replace
  • Shaft bearings are permanently lubricated
  • Order air set separately
Indexed ball-and-socket design with vibration pad lets you adjust the mixer position and achieve various process results you want. Cast aluminum housing.
738,000.00 - ₹826,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Impeller Size (in) Max Speed (rpm) Max Torque (in-oz)
Availability Pricing
Neptune Heavy duty, gear-drive mixer; 1/2-hp, 36"L shaft, 8" impeller 5031100 Mfr # JG-3.4-36-8
COMPARE
8 350 1440
₹738,000.00INR / Each
Neptune Heavy duty, gear-drive mixer; 1/2-hp, 48"L shaft, 10" impeller 5031104 Mfr # JG-3.4-10
COMPARE
10 350 1440
₹826,000.00INR / Each