Nichiryo LE Single-Channel Adjustable-Volume Pipettors

  • Light weight—only 3 oz—reduces hand fatigue
  • User-friendly design
  • Excellent performance for the price
Nichiryo LE pipettors have been designed to lighten your stress during long hours of daily work. The design features an enlarged, streamlined grip that can be kept firmly in hand without much effort. The push button is also enlarged and incorporates a free-spinning button top to eliminate the need to lock the dial after setting the volume. The tip ejector is one-touch attachable and detachable.
39,600.00INR / Each

5 variations of this product are available.

See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Volume (mL) Min Volume (mL)
Availability Pricing
Nichiryo 00-NLE-10 Adjustab-Volume Pipettor, 0.5 to 10 uL; Each 0785903 Mfr # 00-NLE-10
COMPARE
0.01 0.0005
₹39,600.00INR / Each
Nichiryo 00-NLE-20 Adjustab-Volume Pipettor, 2 to 20 uL; Each 0785905 Mfr # 00-NLE-20
COMPARE
0.02 0.002
₹39,600.00INR / Each
Nichiryo 00-NLE-100 Adjustab-Volume Pipettor, 10 to 100 uL; Each 0785907 Mfr # 00-NLE-100
COMPARE
0.1 0.01
₹39,600.00INR / Each
Nichiryo 00-NLE-200 Adjustab-Volume Pipettor, 20 to 200 uL; Each 0785909 Mfr # 00-NLE-200
COMPARE
0.2 0.02
₹39,600.00INR / Each
Nichiryo 00-NLE-1000 Adjustable-Volume Pipettor, 100 to 1000 uL; Each 0785911 Mfr # 00-NLE-1000
COMPARE
1 0.1
₹39,600.00INR / Each
Showing 1- 5 of 5

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat