North Sciences Minicore Compact Ultra-Low Temperature Freezer
Minicore freezers are small enough to fit on a tabletop and ensure reliability with cloud-based monitoring
  • Cloud-based monitoring provided by TraceableLive® to securely monitor your critical environments and notify your smartphone, tablet, or PC if anything goes wrong
  • Ideal for COVID-19 vaccines, critical material storage, or anywhere valuable samples need to be stored safely
  • Above industry standard uniformity maintains integrity of samples
  • Green technology uses CFC free natural mix refrigerants
Upright Minicore ultra low temperature freezers provide peace of mind with cloud-based monitoring and are compact enough to fit on your benchtop. They have an above average uniformity at ±2.5°C to ensure that your samples are being stored within proper temperature range. This reliability is enhanced with the inclusion of a Traceable® Ultra-Low Temperature Wi-Fi Data Logger and a 1-year subscription to TraceableLive®. This technology allows you to monitor the freezer remotely from a smartphone, tablet, or PC.
The freezers have an ergonomically designed door handle for easy access to your critical samples. There is also a user-friendly USB port for data download. The refrigerants used in the Minicore are environmentally friendly and the energy usage is low at less than 3A a day.
1,482,000.00 - ₹1,707,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Freezer Capacity (cu ft) Min Temperature - Freezer (° C) Power (VAC)
Availability Pricing
North Sciences Minicore ULT Freezer, -40°C to -86°C, 55 L, with TraceableLive®, 120 V 4426094 Mfr # DF8505-NA
COMPARE
1.9 -86 120
₹1,707,000.00INR / Each
North Sciences Minicore ULT Freezer, -40°C to -86°C, 55 L, with TraceableLive®, 220 V 4426095 Mfr # DF8505-EU
COMPARE
1.9 -86 220
₹1,482,000.00INR / Each