North Sciences/Traceable® Model TEC² Ultra-Low Temperature Freezers
Total sample security and peace of mind with 100% refrigeration redundancy and cloud-based monitoring
  • Smart, self-monitoring freezers have an advanced touch screen control panel
  • Cloud-based monitoring provided by TraceableLive® to securely monitor your critical environments and notify your smartphone, tablet, or PC if anything goes wrong
  • Ideal for COVID-19 vaccines, critical material storage, or anywhere valuable samples need to be stored safely
  • Units have 2 cores, 2 refrigeration systems, and 2 control commands for compressors, enabling one system to maintain temperature if the other is disabled
  • Dual cooling system provides faster pull down and door opening recovery times
The North Sciences Series of Ultra Low Temperature (ULT) Freezers with TEC2 technology have natural, hydrocarbon-based refrigerants to lower GWP (Global Warming Potential), with 100% redundancy provided by two complete and independent refrigeration systems.
Standard ultra-low freezers rely on a cascade refrigeration system. If this system fails, an LN2 or CO2 backup can only sustain temperatures for approximately 7 hours. With TEC2 technology, there is no need to move your samples in a rush or worry about maintenance, refilling, or supervision of LN2 or CO2.
Maintaining temperature and uniformity is important but being aware of external conditions and forces that have the potential to influence the freezer's performance makes the TEC2 smart and situationally aware, for complete performance monitoring and total sample security. It monitors everything from inside out, 24/7. The freezers are integrated with a Traceable® data logger, including a one-year subscription to TraceableLIVE® cloud-based control and monitoring services.
3,213,000.00 - ₹4,503,000.00INR / Each
9 variations of this product are available.
See All Product Options Below
9 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Freezer Capacity (cu ft) Power (Hz) Power (VAC)
Availability Pricing
North Sciences/Traceable Model TEC² ULT Freezer, -40°C to -86°C, 425 L, with TraceableLive®, 120 V/60 Hz 4426085 Mfr # TEC8515-NA
COMPARE
15 60 120
₹3,402,000.00INR / Each
North Sciences/Traceable Model TEC² ULT Freezer, -40°C to -86°C, 425 L, with TraceableLive®, 220 V/50 Hz 4426086 Mfr # TEC8515-EU
COMPARE
15 50 220
₹3,213,000.00INR / Each
North Sciences/Traceable Model TEC² ULT Freezer, -40°C to -86°C, 588 L, with TraceableLive®, 120 V/60 Hz 4426087 Mfr # TEC8520-NA
COMPARE
21 60 120
₹3,502,000.00INR / Each
North Sciences/Traceable Model TEC² ULT Freezer, -40°C to -86°C, 588 L, with TraceableLive®, 220 V/50 Hz 4426088 Mfr # TEC8520-EU
COMPARE
21 50 220
₹3,269,000.00INR / Each
North Sciences/Traceable Model TEC² ULT Freezer, -40°C to -86°C, 728 L, with TraceableLive®, 120 V/60 Hz 4426089 Mfr # TEC8525-NA
COMPARE
26 60 120
₹4,118,000.00INR / Each
North Sciences/Traceable Model TEC² ULT Freezer, -40°C to -86°C, 728 L, with TraceableLive®, 208-220 V/60 Hz 4426090 Mfr # TEC8525-NA-2
COMPARE
26 60 220
₹4,118,000.00INR / Each
North Sciences/Traceable Model TEC² ULT Freezer, -40°C to -86°C, 728 L, with TraceableLive®, 220 V/50 Hz 4426091 Mfr # TEC8525-EU
COMPARE
26 50 220
₹3,829,000.00INR / Each
North Sciences/Traceable Model TEC² ULT Freezer, -40°C to -86°C, 838 L, with TraceableLive®, 208-220 V/60 Hz 4426092 Mfr # TEC8530-NA
COMPARE
30 60 220
₹4,503,000.00INR / Each
North Sciences/Traceable Model TEC² ULT Freezer, -40°C to -86°C, 838 L, with TraceableLive®, 208-220 V/50 Hz 4426093 Mfr # TEC8530-EU
COMPARE
30 50 220
₹4,176,000.00INR / Each
Showing 1- 9 of 9