Oakton® 30 Series Waterproof Pocket Testers
Durable pocket meters for the most challenging routine applications
  • Replaceable, double-junction electrode sensors for highly caustic, volatile or hazardous samples
  • Easy to operate Three-button system
  • IP67-rated housing sits flat on the table and floats in water
Our most capable series of pocket testers, the Oakton 30 Series include pHTestr 30, ORPTestr 30, and PCTSTestr 30. Replaceable and double junction sensors combine to provide durability and longevity against challenging substance properties in both lab and field applications. Waterproof and designed to float in water, they are ideal for field-based testing environments – with a convenient loop for security line attachment.
30 series pocket meters include a replaceable double-junction electrode sensor that is designed for more aggressive applications, including wastewater and solutions containing sulfides, heavy metals, and TRIS. The electrode sensor offers a clog-resistant Kynar® junction and a recessed, glass pH bulb to protect against breakage and allow for easy maintenance and replacement. A Valox sensor housing resists most chemicals.
An easy-to-use three-button system allows for technician level use and measurement. Up to 3 calibration points with auto-buffer recognition at both USA and NIST standard buffers. Additional features include an automatic temperature compensation (ATC), hold function, auto-off function, self-diagnostic error messages, and a three-segment battery-life indicator.
17,300.00 - ₹39,100.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
Waterproof pH Testr 30 Pocket Tester
₹17,300.00INR / Each
COMPARE
Double Junction Waterproof ORP Pocket Tester
₹19,100.00INR / Each
COMPARE
Waterproof Multiparameter Pocket Tester
₹39,100.00INR / Each