Oakton® Benchtop CON 700 Meter
An affordable way to take consistent, accurate conductivity measurements in your laboratory!
  • Compact size frees up bench space
  • Large brightly lit display allows for easy reading of measured values
  • Easily toggle between conductivity and TDS at the touch of a button!
CON 700 meter reads 0 to 200 mS across five ranges—autoranging feature automatically gives the best resolution for your measurements. Calibrate at up to five conductivity points (one per range) for improved measurement accuracy. Adjustable temperature coefficient provides additional accuracy. Cell constant is selectable for very high or low range measurements - meter is flexible to your application. Selectable manual or automatic temperature compensation gives greater precision.
Meter is encased in a compact 6-1/8" L x 6-7/8" W x 2-3/4" (15.5 x 17.5 x 6.9 cm) housing which takes up a minimum of bench space-nearly 40% less space than comparable meters. Water-resistant membrane keypad and convenient pullout reference guide provide useful features for the lab. Oversized display measures 3-1⁄4" x 2-7⁄16" (8.3 x 6.2 cm) and is brightly lit for easy viewing of measurements. Display shows conductivity or TDS and temperature and has a “ready” indicator and Hold function.
Additional meters features include, electrode stand that is switchable to either side of the meter, and °C temperature measurement.
76,000.00 - ₹80,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Conductivity Resolution Max Conductivity Min Conductivity
Availability Pricing
COMPARE
0.01 or 0.1 μS; 0.001, 0.01, or 0.1 mS 200 mS 0.01 uS
₹76,000.00INR / Each
COMPARE
0.01 or 0.1 μS; 0.001, 0.01, or 0.1 mS 200.0 mS 0.02 uS
₹80,000.00INR / Each