Environmental Express/ Oakton® pH Epoxy-Body Electrodes with ATC
Environmental Express/ Oakton offers a variety of pH electrodes for your general purpose and speciality laboratory applications
  • Choose single-junction electrodes to measure pH in clean aqueous samples
  • Choose double-junction electrodes for use with samples that may be contaminating with particulates, heavy metals or organic compounds
Low-maintainance epoxy-bodied electrodes provide the durability for field work. Refillable electrodes provide greater accuracy and faster response for more critical analytical work.
19,000.00 - ₹28,500.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
ATC Connector Electrode Type
Availability Pricing
COMPARE
6-pin waterproof Single junction
₹19,000.00INR / Each
COMPARE
6-pin waterproof Double junction
₹28,500.00INR / Each