Ohaus Rocking and Waving Shakers
Simple tilt angle and speed adjustments provide reliable rocking and waving
  • Intuitive touchpad controls
  • Easily adjust tilt angle in 1° increments
  • Speed ramping feature slowly increases speed to avoid splashing
These rocking and waving shakers are designed for diverse laboratory applications, offering controlled motion for effective mixing. The rocking shakers provide seesaw-like movement, while the waving shakers deliver a gentle 3D wave motion.
You can easily adjust tilt angle in 1° increments and speed down to 1 rpm with the digital models, even while in operation. Intuitive touchpad controls make setup straightforward. Electronic control ensures reproducible results, with a speed ramping feature to prevent splashing.
All models include a non-slip mat while many models have stacking tray options for increased capacity.
194,000.00 - ₹348,000.00INR / Each
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity Max Speed (rpm) Power (VAC)
Availability Pricing
Ohaus SHRK04DG Rocking Digital Shaker, 1 Tier Platform 5302685 Mfr # SHRK04DG
COMPARE
10 lb (4.5 kg) 50 120
₹236,000.00INR / Each
Ohaus SHRK07AL1 Rocking Analog Shaker, 1 Tier Platform 5302686 Mfr # SHRK07AL1
COMPARE
16 lb (7.3 kg) 75 120
₹194,000.00INR / Each
Ohaus SHRK07AL2 Rocking Analog Shaker, 2 Tier Platform 5302687 Mfr # SHRK07AL2
COMPARE
16 lb (7.3 kg) 75 120
₹214,000.00INR / Each
Ohaus SHWV02DG Waving Digital Shaker 5302688 Mfr # SHWV02DG
COMPARE
5 lb (2.3 kg) 30 120
₹348,000.00INR / Each
Ohaus SHWV02AL Waving Analog Shaker 5302689 Mfr # SHWV02AL
COMPARE
5 lb (2.3 kg) 75 120
₹276,000.00INR / Each