Oil-less Diaphragm Air Compressors
Virtually maintenance-free
  • Offers oil-free, cool air, even at maximum duty — air entering the pump does not go near the motor to pick up heat.
  • Ideal for light duty applications
Diaphragm Air Compressors offer oil-free, cool air, even at maximum duty—air entering the pump does not go near the motor to pick up heat.
135,000.00 - ₹141,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Free Air Capacity (CFM) Max Pressure (PSI) Power (VAC)
Availability Pricing
Gast DOA-P501-BN Oilless Air Compressor, Diaphragm compressor pump, 1.1 cfm, 220/240 VAC 0705415 Mfr # DOA-P501-BN
COMPARE
1.1 60 220 / 240
₹141,000.00INR / Each
Gast DOA-P501-FD Oilless Air Compressor, Diaphragm compressor pump, 1.9 cfm, 220/240 VAC 0705423 Mfr # DOA-P501-FD
COMPARE
1.9 60 220 / 240
₹135,000.00INR / Each