PCRmax® Human Viral Pathogen Detection Kits for qPCR
Crucial tool for fast and accurate detection of dangerous human viral pathogens
  • No need to refrigerate—all contents are lyophilized for simple transport and storage
  • Positive copy number standard curve provides accurate quantification
  • Exceptional value at 150 tests per kit
These detection kits utilize the sensitivity and speed of qPCR to get the most accurate data as quickly as possible. Each kit is specifically designed by our bioinformatics team to ensure the broadest possible detection profile and detection of all relevant strains and subtypes. All test kits are validated in house on multiple qPCR platforms to ensure cross platform functionality. Kits come with sufficient controls to reduce the chances of making the wrong call. Kits can also by generated specifically for your test of interest. If the kit is not in our database, we can easily make the kit you wish if sequence data is available. Kits conform to the latest safety directives including European CE requirements.
Order a complete kit with mastermix or a kit without mastermix and use your own mastermix.
100 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Mastermix Included Test Type Description
Availability Pricing
COMPARE
No RNA H1N1 Influenza, without mastermix
₹238,000.00INR / Each
COMPARE
Yes RNA H1N1 Influenza, with mastermix
₹339,000.00INR / Each
COMPARE
No DNA Adenovirus type C, without mastermix
₹164,000.00INR / Each
COMPARE
Yes DNA Adenovirus type C, with mastermix
₹209,000.00INR / Each
COMPARE
No RNA Coronavirus genomes (all Group 1), without mastermix
₹164,000.00INR / Each
COMPARE
Yes RNA Coronavirus genomes (all Group 1), with mastermix
₹214,000.00INR / Each
COMPARE
No RNA Coronavirus genomes (all Group 2), without mastermix
₹164,000.00INR / Each
COMPARE
Yes RNA Coronavirus genomes (all Group 2), with mastermix
₹214,000.00INR / Each
COMPARE
No RNA Human enterovirus (all species), without mastermix
₹201,000.00INR / Each
COMPARE
Yes RNA Human enterovirus (all species), with mastermix
₹256,000.00INR / Each
COMPARE
No DNA BK Virus, without mastermix
₹164,000.00INR / Each
COMPARE
Yes DNA BK Virus, with mastermix
₹209,000.00INR / Each
COMPARE
No RNA Chikungunya Virus, without mastermix
₹164,000.00INR / Each
COMPARE
Yes RNA Chikungunya Virus, with mastermix
₹214,000.00INR / Each
COMPARE
No DNA Cytomegalovirus (HHV5), without mastermix
₹164,000.00INR / Each
COMPARE
Yes DNA Cytomegalovirus (HHV5), with mastermix
₹209,000.00INR / Each
COMPARE
No RNA Dengue Virus, without mastermix
₹164,000.00INR / Each
COMPARE
Yes RNA Dengue Virus, with mastermix
₹228,000.00INR / Each
COMPARE
No RNA Dengue Virus type 3, without mastermix
₹164,000.00INR / Each
COMPARE
Yes RNA Dengue Virus type 3, with mastermix
₹214,000.00INR / Each
Showing 1 - 20 of 100