Pfeiffer Magnetically Coupled Rotary Vane Vacuum Pumps
Sealed head, means no oil leaks
  • High pumping speeds
  • Safety valve seals the pump during power loss
  • Easy to service, low maintenance
Magnetically coupled drive shaft eliminates oil leaks. Dual mode pump with gas ballast allows for vacuum settings for many different applications.
Call For Price
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Free-Air Capacity (CFM) Max Vacuum (torr) Power (VAC)
Availability Pricing
Pfeiffer PK-D63-712 Magnetically couple rotary vane, 14 CFM, 230 VAC 7950045 Mfr # PK-D63-712
COMPARE
14.1 0.0037 230