Plas-Labs Compact Glove Boxes/Balance Chambers
Enables safe handling of dangerous materials or microorganisms
  • Prevent moisture absorption while handling materials
  • Provide personal protection from harmful substances
Gloved containment chambers offer you a safe, clean workspace that's ideal for enclosing balances or scales. HEPA and organic vapor filter safety valves protect you from particulates and harmful vapors. Four gas purging valves enable operation of weighing and transfer chambers. Top section is easily removable for equipment transfer.
Call For Price
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Chamber Depth (in) Chamber Height (in) Power (VAC)
Availability Pricing
Plas-Labs 830-ABC/EXP Weighing Chamber / Compact Glove Box, 11 cu ft; 230v 3475242 Mfr # 830-ABC/EXP
COMPARE
28 23 220
Call For Price
Plas-Labs 830-ABB/EXP Weighing Chamber / Compact Glove Box, 17 cu ft; 230v 3475247 Mfr # 830-ABB/EXP
COMPARE
29 30 220