Platinum Crucible Lids
Prohibit contamination at high temperatures up to 2192°F (1200°C)
  • Constructed of durable platinum
Platinum offers oxidation resistance in air and oxygen; and corrosion resistance in acids and melted salts. Lids, crucibles, and tongs sold separately.
315,000.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Material Description
Availability Pricing
Platinum Cover for 15 mL Standard Form Crucibles, 1/ea 1783125 Mfr # GCS15LID
COMPARE
15 Platinum Cover for 15 mL Standard Form Crucibles, 1/ea
Call For Price
Platinum Cover for 40 mL Standard Form Crucibles, 1/ea 1783165 Mfr # GCS40LID
COMPARE
40 Platinum Platinum Cover for 40 mL Standard Form Crucibles, 1/ea
Call For Price
Platinum Cover for 50 mL Standard Form Crucibles, 1/ea 1783175 Mfr # GCS50LID
COMPARE
50 Platinum Platinum Cover for 50 mL Standard Form Crucibles, 1/ea
₹315,000.00INR / Each