PRO Scientific PRO400DEL Digital Benchtop Homogenizer
Homogenize, emulsify, or blend micro- to batch-sized samples
  • Thoroughly mix sample volumes from 0.03 mL to 20 L (5.3 gal.) at up to 23,000 rpm
  • Finely control the speed, digitally, in 100 rpm increments
  • Quickly connect the generator probe that works for your applications
  • Effortlessly adjust height with the motorized stand
This homogenizer is the perfect combination of power and technology with a 1-3/4 hp motor and bright digital display. The speed-control knob allows for 100 rpm resolution, as opposed to 1000 rpm increments with most homogenizers. The motorized test stand allows for smooth control of motor height and adds 11" for compatibility with taller vessels.
Choose a generator probe based on the sample volume and dispersion type (order separately). The flat-bottom probe is for small volumes, the open-slotted is for blending low-viscosity fluids or soft tissues, and the saw-tooth creates enough shear to handle tough tissues. The Multi-Gen system offers low-cost repeat homogenization without the risk of cross-contamination. Clean and sterilize the 316 stainless steel and PTFE Multi-Gen probes.
Call For Price
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Sample Size (mL) Min Sample Size (mL) Power (VAC)
Availability Pricing
PRO Scientific PRO400DEL Digital Benchtop Homogenizer and Motorized Stand; 120V 0475140 Mfr # 01-01400DSEL
COMPARE
20000 0.03 120
Call For Price
PRO Scientific PRO400DEL Digital Benchtop Homogenizer and Motorized Stand; 220V 0475145 Mfr # 01-02400DSEL
COMPARE
20000 0.03 220