Ralston Instruments Hose and Adapter Kits
Adapters have a unique sealing system that makes them excellent for both high and low pressure calibration.
  • ABS case with foam cut-outs makes it easy to see if any adapters are missing
  • Adapters are available in kits or individually.
Kits take the guesswork out of selecting adapters by providing a selection of the most popular combinations of hoses and adapters necessary to connect to any pressure transmitter, gauge, hand pump, manifold or nitrogen source. Use the individual hoses and fittings to connect to any device you will be calibrating.
3,600.00 - ₹3,900.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Max Pressure Min Pressure Description
Availability Pricing
Ralston Instruments QTHA-1MB0 1/8" male NPT x male QT, no check-valve, brass 6897676 Mfr # QTHA-1MB0
₹3,900.00INR / Each
COMPARE
5000 psi (345 bar) 0 psi (0 bar) 1/8" male NPT x male QT, no check-valve, brass
Ships by May 04
₹3,900.00INR / Each
Ralston Instruments QTHA-2MBO 1/4" male NPT x male QT, no check-valve, brass 6897677 Mfr # QTHA-2MB0
₹3,600.00INR / Each
COMPARE
5000 psi (345 bar) 0 psi (0 bar) 1/4" male NPT x male QT, no check-valve, brass
Ships by May 14
₹3,600.00INR / Each