Roscid Technologies HDR200 Humidity and Temperature Transmitter Accessories

For use with the HDR200 series transmitters
  • Sample chamber and fitting help improve measurements
The HDR200 series is a family of humidity probes and electronic modules that offer % RH and dew point measurements with NIST traceable calibration.
Call For Price

2 variations of this product are available.

See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Color Compatible With Description
Availability Pricing
Roscid Technologies HDR200 Sample Chamber with 1/4" Tube Fittings 7893041 Mfr # HDR200-SC
COMPARE
Black HDR200 transmitters HDR200 Sample Chamber with 1/4" Tube Fittings
Call For Price
Roscid Technologies 1/2" Bore-through Fitting for HDR200 7893042 Mfr # HDR200-1/2 fttg
COMPARE
Black HDR200 transmitters 1/2" Bore-through Fitting for HDR200
Call For Price
Showing 1- 2 of 2

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat