SalvisLab Thermocenter Mechanical Convection Ovens
New SalvisTEQ touch screen controller provides dynamic control behavior, shorter heat-up times, and higher precision
  • Predictive controller monitors what it has done, where it is, and where it wants to go in order to achieve the outcome in the most efficient manner
  • SalvisTEQ is Class 3.3 safety rated with lower and upper temperature limit protection
  • Stainless steel interior and shelves resist aggressive substances
Short heating and precise temperature control make these ovens ideal for a variety of heating and drying tasks. High-quality insulation and airtight design prevent heat loss — these ovens heat your samples, not your lab. Ovens feature an oversized heat ring and adjustable-speed fan to ensure excellent uniformity. In addition to cool-touch operation, these ovens have an insulated silicone door seal that prevents the escape of harmful fumes. The heater and blower stop as soon as you open the oven door.
All-new SalvisTEQ controller has touch screen operation that is clearly laid out and intuitive to operate. Controller features real-time temperature monitoring during the heating-up phase, set point temperature, gradient and actual temperature. The predictive nature of the controller continually monitors oven operation to ensure it operates in the most efficient manner.
Fan control allows fan speed to be adjusted between 40/60 and 100%. Programmable timer let's you to set start time as well as hold time at a desired temperature and simple start/stop button. Other features include self-check for hardware and software issues on each boot, real-time clock, simple switching between °F and °C, 20 MB of internal storage, acoustic and visual alarms, and standard USB connection.
Call For Price
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (cu ft) Power (VAC) Description
Availability Pricing
SalvisLab Thermocenter TC40 Oven, 40 L capacity, 115 VAC 5240301 Mfr # 31A04307
COMPARE
1.4 115 Oven, 40 L capacity, 115 VAC
Call For Price
SalvisLab Thermocenter TC40 Oven, 40 L capacity, 220 VAC 5240306 Mfr # 31A04200
COMPARE
1.4 220 Oven, 40 L capacity, 220 VAC
Call For Price
SalvisLab Thermocenter TC100 Oven, 100 L capacity, 115 VAC 5240311 Mfr # 31A04308
COMPARE
3.5 115 Oven, 100 L capacity, 115 VAC
Call For Price
SalvisLab Thermocenter TC100 Oven, 100 L capacity, 220 VAC 5240316 Mfr # 31A04201
COMPARE
3.5 220 Oven, 100 L capacity, 220 VAC
Call For Price
SalvisLab Thermocenter TC160 Oven, 160 L capacity, 115 VAC 5240321 Mfr # 31A04302
COMPARE
5.6 115 Oven, 160 L capacity, 115 VAC
Call For Price
SalvisLab Thermocenter TC160 Oven, 160 L capacity, 220 VAC 5240326 Mfr # 31A04202
COMPARE
5.6 220 Oven, 160 L capacity, 220 VAC
Call For Price
SalvisLab Thermocenter TC240 Oven, 240 L capacity, 115 VAC 5240331 Mfr # 31A04303
COMPARE
8.4 115 Oven, 240 L capacity, 115 VAC
Call For Price
SalvisLab Thermocenter TC400 Oven, 400 L capacity, 115 VAC 5240341 Mfr # 31A04304
COMPARE
14.1 115 Oven, 400 L capacity, 115 VAC
Call For Price
Showing 1- 8 of 8