Seward Stomacher Sterile Blending Bags
Eliminates the risk of crosss contamination
  • 1 year warranty
Choose from a variety of bags to use with your lab blender. Blending bags are made from high quality food grade nontoxic virgin LDPE granule, extruded to 65µm film thickness. All blender bags are irradiated sterile. A certificate or irridiation is available upon request.
Call For Price
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Bag Capacity (L) Bag Capacity (mL)
Availability Pricing
Seward BA6040 Stomacher® 80 Biomaster Standard Bags, Square Bottom; 2000/CS 3685091 Mfr # BA6040
COMPARE
0.08 80
Call For Price
Seward BA6141 Stomacher® 400 Circulator Standard Bags, Round Bottom; 500/CS 3685093 Mfr # BA6141
COMPARE
0.4 400
Call For Price
Seward BA6041 Stomacher® 400 Circulator Standard Bags, Square Bottom; 500/CS 3685097 Mfr # BA6041
COMPARE
0.4 400
Call For Price
Seward BA6040/STR Stomacher® 80 Biomaster Strainer Bags, Square Bottom; 200/PK 3686000 Mfr # BA6040/STR
COMPARE
0.08 80
Call For Price
Seward   Stomacher® 400 Circulator Strainer Bags, Sterile, 80-400 mL; 200/PK 3686001 Mfr # BA6141/STR
COMPARE
0.4 400