Spectra/Por 1 Regenerated Cellulose (RC) Dialysis Tubing
Get gentle separation and purification of solutes
  • Available in a variety of diameters and flat widths
  • MWCO from 6000 to 8000 daltons
The RC Dialysis tubing is packaged dry to provide you with easy handling and storage. Available in 15 and 30 meter rolls to provide better economy for applications requiring larger amounts of tubing.
Most of the dry tubing rolls contain glycerin as a humectant which is easily removed by soaking 30 min in water.
70,300.00 - ₹160,000.00INR / Box of 1
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Diameter (mm) Flat Width (mm) Max Molecular Weight Cut Off (Mwco) (Dalton)
Availability Pricing
Spectra Por 1 RC Dialysis Tubing, 6-8 kD MWCO, 10mm Flat Width, 6.4mm dia; 50' L 0289070 Mfr # 132645
COMPARE
6.4 10 8000
₹70,300.00INR / Box of 1
Spectra Por 1 RC Dialysis Tubing, 6-8 kD MWCO, 23mm Flat Width, 14.6mm dia; 100' L 0290010 Mfr # 132650
COMPARE
14.6 23 8000
₹71,000.00INR / Box of 1
Spectra Por 1 RC Dialysis Tubing, 6-8 kD MWCO, 32mm Flat Width, 20.4mm dia; 100' L 0290011 Mfr # 132655
COMPARE
20.4 32 8000
₹71,000.00INR / Box of 1
Spectra Por 1 RC Dialysis Tubing, 6-8 kD MWCO, 40mm Flat Width, 25.5mm dia; 100' L 0290012 Mfr # 132660
COMPARE
25.5 40 8000
₹76,400.00INR / Box of 1
Spectra Por 1 RC Dialysis Tubing, 6-8 kD MWCO, 50mm Flat Width, 31.8mm dia; 100' L 0290013 Mfr # 132665
COMPARE
31.8 50 8000
₹89,000.00INR / Box of 1
Spectra Por 1 RC Dialysis Tubing, 6-8 kD MWCO, 100mm Flat Width, 63.7mm dia; 50' L 0290014 Mfr # 132670
COMPARE
63.7 100 8000
₹106,000.00INR / Box of 1
Spectra Por 1 RC Dialysis Tubing, 6-8 kD MWCO, 120mm Flat Width, 76.4mm dia; 50' L 0290015 Mfr # 132675
COMPARE
76.4 120 8000
₹160,000.00INR / Box of 1
Showing 1- 7 of 7