Cole-Parmer® Accessories for Cole-Parmer® MP-100, MP-200, MP-400, and MP-800 Series Melting Point Apparatuses

Accessories for your melting point apparatus
2,600.00 - ₹76,500.00INR / Pkg of 100

7 variations of this product are available.

See All Product Options Below
7 PRODUCT OPTIONS
Compareitem specifications
Compatible With Description
Availability Pricing
COMPARE
Melting point apparatus 03011-49 and colony counters 14212-04 and 14212-06 Printer for melting point apparatus 03011-49 and colony counters 14212-04 and 14212-06
₹76,500.00INR / Each
COMPARE
Analog melting point apparatus 03013-60, 03013-61 Spirit-Filled Thermometer, 50 to 250 °C
Call For Price
COMPARE
03011-40, 03011-41, 03011-45, 03011-47, 03011-49, 03013-60, 03013-61 Glass Capillary Tubes, Open Both Ends; 100/PK
₹4,000.00INR / Pkg of 100
COMPARE
03011-40, 03011-41, 03011-45, 03011-47, 03011-49, 03013-02, 03013-03, 03013-60, 03013-61 Glass Capillary Tubes, Open One End; 100/PK
Call For Price
COMPARE
03011-40, 03011-41, 03011-45, 03011-47, 03011-49, 03013-02, 03013-03, 03013-60, 03013-61 Glass Capillary Tubes, Closed at Both Ends; 100/PK
Call For Price
COMPARE
03013-60, 03013-61 Brass Cooling Plug for Analog Melting Point Apparatus
₹2,600.00INR / Each
COMPARE
03013-02 and 03013-03 Melting Point Calibration Kit
Call For Price
Showing 1- 7 of 7

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat