Thermo Scientific Bantam Deionizer Cartridges
  • Selectable cartridges to best meet your application
Disposable cartridges are easily replaced by turning a thumbscrew.
36,900.00 - ₹87,400.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Description Capacity (grains)
Availability Pricing
Thermo Scientific Barnstead D0760 Deionization Cartridge, anion removal 0150516 Mfr # D0760
COMPARE
Deionization Cartridge, anion removal 1680 as Sodium Chloride
₹87,400.00INR / Each
Thermo Scientific Barnstead D0815 Deionization Cartridge, cation removal 0150518 Mfr # D0815
COMPARE
Deionization Cartridge, cation removal 3170 as Sodium Chloride
₹36,900.00INR / Each