Thermo Scientific™ Nunc™ Disposable Inoculating Loops and Needles
Color-coded for quick and easy identification
  • Radiation sterilized—no need for flame sterilization
  • Hydrophilic treatment during manufacturing process
  • Liquid adherence properties that prevent accidental discharge of droplets
These sterile, polystyrene loops and needles are calibrated using a standard urea solution. A Thermo Scientific™ Nunc™ calibration certificate can be obtained upon request. With no need to flame, these loops and needles save you time. The risk of contamination is eliminated since these instruments are disposable. The surface of each loop and needle receives a special hydrophilic treatment to enhance the adhesion of droplets and prevent accidental discharge. Loops are color coded for easy identification.
66,200.00 - ₹74,900.00INR / Pkg of 1000
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Color Handle Type Description
Availability Pricing
Thermo Scientific Nunc Disposable Inoculating Loop, Polystyrene, Sterile, Blue, 10 µL; 1000/Pk 1421300 Mfr # 251586
COMPARE
Blue Loop Disposable Inoculating Loop, Polystyrene, Sterile, Blue, 10 µL; 1000/Pk
₹74,900.00INR / Pkg of 1000
Thermo Scientific Nunc Disposable Inoculating Loop, Polystyrene, Sterile, Blue, 10 µL; 600/Pk 1421301 Mfr # 254437
COMPARE
Blue Loop Disposable Inoculating Loop, Polystyrene, Sterile, Blue, 10 µL; 600/Pk
₹66,200.00INR / Pkg of 600
Thermo Scientific Nunc Disposable Inoculating Loop, Polystyrene, Sterile, Clear, 1 µL; 1000/Pk 1421310 Mfr # 253287
COMPARE
Clear Loop Disposable Inoculating Loop, Polystyrene, Sterile, Clear, 1 µL; 1000/Pk
₹74,900.00INR / Pkg of 1000
Thermo Scientific Nunc Disposable Inoculating Loop, Polystyrene, Sterile, Clear, 1 µL; 600/Pk 1421311 Mfr # 254410
COMPARE
Clear Loop Disposable Inoculating Loop, Polystyrene, Sterile, Clear, 1 µL; 600/Pk
₹66,200.00INR / Pkg of 600
Thermo Scientific Nunc Disposable Inoculating Needle, Polystyrene, Sterile, Yellow; 600/Pk 1421321 Mfr # 254399
COMPARE
Yellow Needle Disposable Inoculating Needle, Polystyrene, Sterile, Yellow; 600/Pk
₹66,200.00INR / Pkg of 600