Thermo Scientific™ Nunc™ Sterile Non-Treated Cell Culture Flasks
Non-treated surface offers low cell attachment
  • Certified nonpyrogenic and sterile
  • White caps for easy identification
  • The filter cap ensures constant airflow while minimizing chances of contamination
These sterile culture flasks are used for unique cell growth applications that do not require a Nunclon™ Delta surface treatment. Untreated surface offers low attachment. Clear, uniform design provides excellent optical quality, making it a suitable choice for microscopy. Flasks have a white cap for easy identification. All models are certified nonpyrogenic and sterile.
TripleFlask™ model 01928-42 offers a unique three-parallel growth surface that provides a total culture area of 500 cm2 - ideal for scale-up.
EasyFlask™ models 01928-70, -72, and -74 feature an ergonomic design requiring only a 1/3 turn to open and close the cap.
95,300.00 - ₹223,000.00INR / Case of 32
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Culture Area (cm²) Working Volume (mL) Description
Availability Pricing
Thermo Scientific Nunc TripleFlask™ Cell Culture Flask, Polystyrene, Sterile, Non-Treated, Straight Neck, 500 cm2; 32/Cs 0192842 Mfr # 132903
COMPARE
500 200 TripleFlask™ Cell Culture Flask, Straight Neck, 500 cm2; 32/Cs
₹223,000.00INR / Case of 32
Thermo Scientific Nunc EasyFlask™ Cell Culture Flask, Polystyrene, Sterile, Non-Treated, Angled Neck, 25 cm2; 200/Cs 0192870 Mfr # 169900
COMPARE
25 7 EasyFlask™ Cell Culture Flask, Angled Neck, 25 cm2; 200/Cs
₹187,000.00INR / Case of 200
Thermo Scientific Nunc EasyFlask™ Cell Culture Flask, Polystyrene, Sterile, Non-Treated, Angled Neck, 75 cm2; 100/Cs 0192872 Mfr # 156800
COMPARE
75 30 EasyFlask™ Cell Culture Flask, Angled Neck, 75 cm2; 100/Cs
₹163,000.00INR / Case of 100
Thermo Scientific Nunc EasyFlask™ Cell Culture Flask, Polystyrene, Sterile, Non-Treated, Angled Neck, 175 cm2; 30/Cs 0192874 Mfr # 159926
COMPARE
175 55 EasyFlask™ Cell Culture Flask, Angled Neck, 175 cm2; 30/Cs
₹95,300.00INR / Case of 30