Thermo Scientific™ SureSTART™ Autosampler Vials
Certified performance vials are quality tested to deliver the reproducibility you need
  • 2-mL size is ideal for chromatography applications
  • Decrease your risk of failed analysis and sample reruns
  • 100% traceable through the manufacturing process
Meet your certification and documentation needs with the Thermo Scientific™ SureSTART™ specification certified autosampler vials. The unique threaded crown design of these glass vials ensures a secure seal with just a half-turn of the cap (caps sold separately). Precision formed neck and crimp cap profile are compatible with robotic arm autosampler trays. Vials with write-on patches have graduations at 0.5, 1.0, and 1.5 mL.
Choose from Level 2 or 3 compliance. Level 2 products have very low metal contents, extractables, and leachables to ensure that they conform to the high standards you need to meet your regulatory requirements. Level 3 products are packed in clean rooms to protect against environmental factors. They contain the lowest levels of metals, extractables, and leachables to ensure consistent, reproducible sensitivity and selectivity for the most challenging analytes.
8,900.00 - ₹11,500.00INR / Pkg of 100
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Level Tube Color Volume (mL)
Availability Pricing
Thermo Scientific SureSTART™ AVCS™ Autosampler Screw Top Vial, Level 3, Short Thread, Clear Glass, 2 mL; 100/PK 9881000 Mfr # 6PSV9-1P
₹8,900.00INR / Pkg of 100
COMPARE
3 Clear 2
Ships by April 21
₹8,900.00INR / Pkg of 100
Thermo Scientific SureSTART™ AVCS™ Autosampler Screw Top Vial, Level 2, Short Thread, Amber Glass, 2 mL; 100/PK 9881002 Mfr # 6ASV9-2P
₹11,500.00INR / Pkg of 100
COMPARE
2 Amber 2
Ships by May 18
₹11,500.00INR / Pkg of 100