Thermo Scientific™ Target DP™ Vial Caps with Septa
A variety of septa materials to match your equipment and application
  • Prevent septa push through and over- and under-tightening
  • Easy-on, easy-off convenience with just one turn
Get the assurance of a crimp seal with the time-saving convenience of a screw cap. The unique threaded crown design ensures a secure seal with just a half-turn of the cap.
19,200.00 - ₹20,400.00INR / Pkg of 100
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Thermo Scientific C-4000-53B Caps with Septa; red PTFE/white silicone/red PTFE 9881022 Mfr # 6PSC9TST
COMPARE
Caps with Septa; red PTFE/white silicone/red PTFE
₹19,200.00INR / Pkg of 100
Thermo Scientific C-4000-55B Caps with Septa; pre-slit blue PTFE/white silicone 9881024 Mfr # 6PSC9STS1
COMPARE
Caps with Septa; pre-slit blue PTFE/white silicone
₹20,400.00INR / Pkg of 100