Thermo Scientific Tube Revolver
Interchangeable paddles allow for maximum flexibility in tube choice
  • Small footprint
  • Paddles are adjustable from 0 to 90° to optimize the mixing angle
  • Set in “oscillating” mode to achieve more effective mixing, oscillation occurs every 40 degrees in the rotation cycle
  • Variable speed with easy-to-use intuitive controls
  • Maintenance-free motor provides quiet operation
141,000.00 - ₹162,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Speed (rpm) Power (VAC)
Availability Pricing
Thermo Scientific 88881001 Tube Revolver, 100-240 VAC; US Plug 0472516 Mfr # 88881001
COMPARE
40 100 to 240
₹141,000.00INR / Each
Thermo Scientific 88881002 Tube Revolver, 100-240 VAC; International Plug 0472517 Mfr # 88881002
COMPARE
40 100 to 240
₹162,000.00INR / Each