ThermoSafe Brands Insulated PUR Shippers
Provide extensive protection for long-range trips
Rigid, reusable, one-piece, three-layer laminate construction provides excellent temperature protection, and exceptional strength and durability. Durable closed-cell polyurethane foam insulated shippers ensure frozen or refrigerated products from moderate and extreme ambient temperature exposure. Flexible polyurethane foam plug seals the top, cushions and insulates contents, and vents dry ice. Shippers withstand crush compression in excess of 5000 lbs. CFC and HCFC free; comply with international clean air regulations.
21,000.00 - ₹39,400.00INR / Pkg of 3
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Inside Height (in) Inside Length (in) Inside Width (in)
Availability Pricing
ThermoSafe E36UPS Insulated Shipper, 2" Wall, Small 9953080 Mfr # E36UPS
COMPARE
7 11 8
₹39,400.00INR / Pkg of 3
ThermoSafe E89UPS Insulated Shipper, 2" Wall, Medium 9953082 Mfr # E89UPS
COMPARE
10 1/4 16 1/4 9 1/4
₹27,600.00INR / Pkg of 2
ThermoSafe E186UPS Insulated Shipper, 2" Wall, Large 9953084 Mfr # E186UPS
COMPARE
12 18 1/2 14 1/2
₹21,000.00INR / Pkg of 1
ThermoSafe E327UPS Insulated Shipper, 2" Wall, Extra large 9953086 Mfr # E327UPS
COMPARE
16 3/4 18 1/2 18 1/4
₹23,900.00INR / Pkg of 1
ThermoSafe E90UPS Insulated Shipper, 3" Wall, Large 9953090 Mfr # E90UPS
COMPARE
13 12 10
₹31,500.00INR / Pkg of 1