ThermoSafe Brands Phase Change Soft Cold Packs
Phase change formula lasts 15% longer that standard gel packs
  • Four formulations from 32 to -10°F (0 to -23°C)
  • Nontoxic and reusable
Keep your sensitive materials at the right temperature. High-tech phase change formula is ideal for products requiring an accurate, consistent temperature — returns to gel state when thawed.
Soft Cold Packs are manufactured with laminated, metallized film exterior to resist punctures and provide a radiant heat barrier.
5,600.00 - ₹9,000.00INR / Case of 24
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Min Temperature (° C) Outside Length (in) Outside Width (in)
Availability Pricing
ThermoSafe 412 Phase Change Cold Pack, Soft, 30F, 12 OZ 0634774 Mfr # 412
COMPARE
-1 6 3/4 4
₹8,400.00INR / Case of 24
ThermoSafe 414 Phase Change Cold Pack, Soft, 30F, 24 oz 0634775 Mfr # 414
COMPARE
-1 7 1/2 6 1/2
₹6,200.00INR / Case of 12
ThermoSafe 426 Phase Change Cold Pack, Soft, 30F, 48 oz 0634776 Mfr # 426
COMPARE
-1 10 1/2 7 3/4
₹5,600.00INR / Case of 6
ThermoSafe 418 Phase Change Cold Pack, Soft, -10F, 12 oz 0634781 Mfr # 418
COMPARE
-23 6 3/4 4
₹9,000.00INR / Case of 24
ThermoSafe 420 Phase Change Cold Pack, Soft, -10F, 24 oz 0634782 Mfr # 420
COMPARE
-23 7 1/2 6 1/2
₹6,900.00INR / Case of 12
ThermoSafe 427 Phase Change Cold Pack, Soft, -10F, 48 oz 0634783 Mfr # 427
COMPARE
-23 10 1/2 7 3/4
₹7,700.00INR / Case of 6