ThermoSafe Envoy Insulated Shippers

The perfect solution for small, one to two day refrigerated shipments
  • Shippers keep products at 36 to 46°F (2 to 8°C) in transit
Multicomponent packing system includes the components necessary to ensure temperature protection for one to two day shipments. Expanded polystyrene (EPS) insulation and refrigerator/freezer packs control the temperature in the shipper.
8,200.00 - ₹12,300.00INR / Each

3 variations of this product are available.

See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
ThermoSafe ERSS1SEU Insluated Summer Packout Shipper, 30 hr, 7 x 5 x 3 in 0373150 Mfr # ERSS1SEU
COMPARE
Insluated Summer Packout Shipper, 30 hr, 7 x 5 x 3 in
₹8,200.00INR / Each
ThermoSafe ERSS1LEU Insluated Summer Packout Shipper, 30 hr, 10.5 x 8.75 x 4 in 0373152 Mfr # ERSS1LEU
COMPARE
Insluated Summer Packout Shipper, 30 hr, 10.5 x 8.75 x 4 in
₹8,900.00INR / Each
ThermoSafe ERSS2LEU Insluated Summer Packout Shipper, 54 hr, 10.5 x 8.75 x 4.5 in 0373156 Mfr # ERSS2LEU
COMPARE
Insluated Summer Packout Shipper, 54 hr, 10.5 x 8.75 x 4.5 in
₹12,300.00INR / Each
Showing 1- 3 of 3

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat