ThermoSafe Lab Mailers

Ship non-hazardous specimens safely
  • Fitted halves cushion tubes for superior protection
  • Lightweight mailers meet postal regulations for nonhazardous materials
Protect your fragile samples with this expanded polystyrene shipper. Light weight minimizes postage costs. Strong white paperboard shipping carton (sold separately) has sure-lock tabs that eliminate taping or tying. Choose from tube mailers, slide mailer, mini mailer, multipurpose mailer, or aluminum shipping can.
13,300.00 - ₹43,200.00INR / Case of 150

5 variations of this product are available.

See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Shipping Weight (oz) Description
Availability Pricing
ThermoSafe 339 Shipper, 5 Tube Mailer, 13 x 103 mm tubes, 150/cs 0372701 Mfr # 339
COMPARE
64 Shipper, 5 Tubes, 13 x 103 mm tubes, 150/cs
₹22,800.00INR / Case of 150
ThermoSafe 356 Shipper, 1 Tube Mailer, 16 x 103 mm tubes, 300/cs 0372702 Mfr # 356
COMPARE
89.6 Shipper, 1 Tube, 16 x 103 mm tubes, 300/cs
₹36,900.00INR / Case of 300
ThermoSafe 341 Shipper, 8 Tube Mailer, 16 x 103mm tubes, 50/cs 0372703 Mfr # 341
COMPARE
89.6 Shipper, 8 Tubes, 16 x 103mm tubes, 50/cs
₹13,300.00INR / Case of 50
ThermoSafe 440 Mini Multipurpose Mailers, 24/cs 0374120 Mfr # 440
COMPARE
94 Mini Multipurpose Mailers, 24/cs
₹21,900.00INR / Case of 24
ThermoSafe 369 Microscope Slide Mailer, holds 10 slides, 300/cs 0375010 Mfr # 369
COMPARE
139 Microscope Slide Mailer, 10 slides, 1" x 3"; 300/cs
₹43,200.00INR / Case of 300
Showing 1- 5 of 5

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat