Traceable® Deluxe Remote-Probe Digital Thermometer with Calibration
10-ft flexible cord makes it easy to reach difficult areas
  • Includes NIST-Traceable certificate from our A2LA accredited (1750.01) calibration laboratory
  • Readings update every second
  • °F/°C switchable ranges
Deluxe remote-probe thermometers feature high/low alarm, MIN/MAX, and HOLD. Magnetic back allows for placement on metal surfaces such as freezers, incubators or water baths. Includes a flip-open stand for use on bench or desktop, and spring fastener for clipping on to any edge.
Traceable® Products have an individually-numbered Traceable® Certificate provided with each unit, that assures accuracy from our ISO/IEC 17025:2017(1750.01) calibration laboratory accredited by A2LA. It indicates traceability of measurements to the SI units through NIST or other recognized national measurement institutes (NMI) that are signatories to the CIPM Mutual Recognition Agreement. Each product goes through an intense vetting process before it can carry the Traceable name, thus saving you both time and money by not having to calibrate separately.
8,600.00 - ₹15,800.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Temperature (° F) Min Temperature (° F) Description
Availability Pricing
Traceable Calibrated Deluxe Remote Probe Thermometer; ±1.0° accuracy (-20 to 100°C) 9020521
₹8,600.00INR / Each
COMPARE
572 -58 Calibrated Deluxe Remote Probe Thermometer; ±1.0° accuracy (-20 to 100°C)
Ships by April 10
₹8,600.00INR / Each
Traceable Calibrated Deluxe Remote Probe Thermometer Ultra; ±0.5°C Accuracy at Tested Points 9876738
₹15,800.00INR / Each
COMPARE
572 -58 Calibrated Deluxe Remote Probe Thermometer Ultra; ±0.5°C Accuracy at Tested Points
Ships by April 10
₹15,800.00INR / Each