Traceable® Ultra-Low Temperature Wi-Fi Data Loggers Compatible with TraceableLIVE® Cloud Service
Monitor critical environments from your mobile devices
  • Connect to your data via smartphone, tablet, or PC using TraceableLIVE wireless capability*
  • View current conditions anywhere—cloud-based interface sends email, text, and push notifications
  • No software required—web, Android™, Apple® iOS data visibility
  • Includes NIST-Traceable certificate from our A2LA accredited (1750.01) calibration laboratory
TraceableLIVE Wireless Technology allows multiple users to stay connected and monitor critical environments 24/7 wherever you go.
These data loggers with Wi-Fi provide a simple, efficient, and reliable way to ensure critical samples are not compromised due to temperature variations. Data loggers measure down to –130°F (–90°C) with ±1.8°F (±0.1°C) accuracy—ideal for ultra-low temperature freezers.
Once setup is complete you can securely monitor parameters 24/7, control your alarms remotely, view your data logging history, run reports in real time, and give access to other team members. No local software is required to use the cloud-based data interface. Get mobile push notifications, emails, or texts for out of range conditions, connectivity interruptions, low-battery condition or if your device is due for calibration. Large tripe-line LCD displays current and Min/Max readings.
Use the included hook-and-loop strip to mount the probe to the inside of a freezer. Micro cable on the probe permits freezer door to close on it. Mount the logger to the outside of the freezer with the included magnetic strip or use the built-in bench stand to place on countertop.
Plus the NIST-traceable precalibration means you get reliable measurements right out of the box, saving you time and money!
Traceable® Products have an individually-numbered Traceable® Certificate provided with each unit, that assures accuracy from our ISO/IEC 17025:2017(1750.01) calibration laboratory accredited by A2LA. It indicates traceability of measurements to the SI units through NIST or other recognized national measurement institutes (NMI) that are signatories to the CIPM Mutual Recognition Agreement. Each product goes through an intense vetting process before it can carry the Traceable name, thus saving you both time and money by not having to calibrate separately.
72,800.00 - ₹84,700.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Min Temperature (° C) Min Temperature (° F) Number of Channels
Availability Pricing
COMPARE
-90 -130 1
₹72,800.00INR / Each
COMPARE
-90 -130 2
₹84,700.00INR / Each