Turbine Propellers, 304 Stainless Steel
Provides airless mixing in deep containers
These turbine propellers mount singularly or with one or more mid-shaft propellers (model 04370-17) for a multi-stage system. Threaded 10 mm dia bore requires shaft 04370-02.
11,900.00 - ₹17,200.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Bore Diameter (in) Material Overall Diameter (in)
Availability Pricing
COMPARE
0.4 Stainless Steel 2.4
₹11,900.00INR / Each
COMPARE
0.4 Stainless Steel 3.9
₹17,200.00INR / Each