Tygon® A-60-F Food and Beverage Tubing
Withstands a wide temperature range from -60 to 135°C (-75 to 275°F)
  • Ideal for general-purpose, laboratory, and single-use applications
  • Free technical application support available to assist with tubing selection
  • Excellent flexural fatigue resistance—great for use in peristaltic pumps
  • Steam clean in place to eliminate frequent tubing replacement
  • Chemically compatible with a variety of cleaning solutions
Tygon® A-60-F tubing will not crack or deteriorate even in extreme temperatures. Featuring high flexural fatigue resistance, tubing is perfect for use in peristaltic pumps often found food and beverage dispensing equipment. It resists kinks, retains its shape, and remains flexible for quick and easy installation.
This high-performance tubing withstands harsh sanitizing solutions and exhibits exceptional acid and alkali resistance. Easily clean tubing using clean-in-place or steam-in-place cleaning and sterilization systems or autoclave up to five cycle times without affecting the service life.
Call For Price
11 variations of this product are available.
See All Product Options Below
11 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Tubing ID (in) Tubing OD (in) Wall Thickness (in)
Availability Pricing
Tygon A-60-F Tubing, 1/16" ID x 3/16" OD; 50 Ft 5010599 Mfr # AAL00003
COMPARE
1/16 3/16 1/16
Call For Price
Tygon A-60-F Tubing, 1/8" ID x 1/4" OD; 50 Ft 5010600 Mfr # AAL00007
COMPARE
1/8 1/4 1/16
Call For Price
Tygon A-60-F Tubing, 3/16" ID x 5/16" OD; 50 Ft 5010601 Mfr # AAL00012
COMPARE
3/16 5/16 1/16
Call For Price
Tygon A-60-F Tubing, 1/4" ID x 3/8" OD; 50 Ft 5010602 Mfr # AAL00017
COMPARE
1/4 3/8 1/16
Call For Price
Tygon A-60-F Tubing, 1/4" ID x 1/2" OD; 50 Ft 5010603 Mfr # AAL00019
COMPARE
1/4 1/2 1/8
Call For Price
Tygon A-60-F Tubing, 5/16" ID x 7/16" OD; 50 Ft 5010604 Mfr # AAL00022
COMPARE
5/16 7/16 1/16
Call For Price
Tygon A-60-F Tubing, 3/8" ID x 1/2" OD; 50 Ft 5010605 Mfr # AAL00027
COMPARE
3/8 1/2 1/16
Call For Price
Tygon A-60-F Tubing, 3/8" ID x 5/8" OD; 50 Ft 5010606 Mfr # AAL00029
COMPARE
3/8 5/8 1/8
Call For Price
Tygon A-60-F Tubing, 1/2" ID x 3/4" OD; 50 Ft 5010607 Mfr # AAL00038
COMPARE
1/2 3/4 1/8
Call For Price
Tygon A-60-F Tubing, 5/8" ID x 7/8" OD; 50 Ft 5010608 Mfr # AAL00046
COMPARE
5/8 7/8 1/8
Call For Price
Tygon A-60-F Tubing, 3/4" ID x 1" OD; 50 Ft 5010609 Mfr # AAL00053
COMPARE
3/4 1 1/8
Call For Price
Showing 1- 11 of 11