Tygon® S3™ B-44-3 Beverage Dispensing Tubing
Excellent nonwetting properties allow complete drainage and easy cleaning
  • Ideal for general-purpose, laboratory, and single-use applications
  • Free technical application support available to assist with tubing selection
  • Minimal impact to the taste or odor of products it transfers
  • Excellent chemical resistance
  • Lightweight and easy to handle
  • Clear tubing provides visual monitoring of flow
  • Phthalate and BPA free
Use the Tygon® S3™ B-44-3 tubing for transferring a wide variety of beverages including soft drinks, fruit juices, and flavored teas. This specially formulated tubing does not affect the taste or odor of transferred products, and the transferred products do not affect the integrity of the tubing such as color, cracking, hardness, tackiness, and flexibility.
The nonwetting properties of the tubing provide complete drainage and allow simple flush cleaning. Lightweight and flexible, the tubing bends to accommodate corners and obstructions and requires minimal fittings for quick and easy installation. Clear tubing allows you to visually monitor the flow, so you can control or adjust the flow during the process.
Call For Price
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Tubing ID (in) Tubing OD (in) Wall Thickness (in)
Availability Pricing
Tygon S3™ B-44-3 Tubing, 1/8" ID x 1/4" OD; 50 Ft 5010669 Mfr # AAB00007
COMPARE
1/8 1/4 1/16
Call For Price
Tygon S3™ B-44-3 Tubing, 3/16" ID x 5/16" OD; 50 Ft 5010670 Mfr # AAB00012
COMPARE
3/16 5/16 1/16
Call For Price
Tygon S3™ B-44-3 Tubing, 1/4" ID x 3/8" OD; 50 Ft 5010671 Mfr # AAB00017
COMPARE
1/4 3/8 1/16
Call For Price
Tygon S3™ B-44-3 Tubing, 3/8" ID x 1/2" OD; 50 Ft 5010672 Mfr # AAB00027
COMPARE
3/8 1/2 1/16
Call For Price
Tygon S3™ B-44-3 Tubing, 1/2" ID x 3/4" OD; 50 Ft 5010673 Mfr # AAB00038
COMPARE
1/2 3/4 1/8
Call For Price
Tygon S3™ B-44-3 Tubing, 5/8" ID x 7/8" OD; 50 Ft 5010674 Mfr # AAB00046
COMPARE
5/8 7/8 1/8
Call For Price
Tygon S3™ B-44-3 Tubing, 3/4" ID x 1" OD; 50 Ft 5010675 Mfr # AAB00053
COMPARE
3/4 1 1/8
Call For Price
Tygon S3™ B-44-3 Tubing, 1" ID x 1-1/4" OD; 50 Ft 5010676 Mfr # AAB00062
COMPARE
1 1 1/4 1/8
Call For Price
Showing 1- 8 of 8