Tygon® S3™ E-3603 Vacuum Tubing
Formulated to resist flex-fatigue and abrasion
  • Bio-based formulation reduces environmental impact
  • Lot-to-lot consistency for reproducible results
  • Easily slips over fittings and grips securely
  • Contains no BPA or phthalates
  • Meets all USP Class VI and FDA food contact criteria
  • Ideal for general-purpose, laboratory, and single-use applications
  • Free technical application support available to assist with tubing selection
Able to handle most organic chemicals found in the lab, the clear and flexible Tygon® S3™ E-3603 is non-oxidizing and non-contaminating. The tubing is less permeable than rubber tubing and features a glassy-smooth inner bore which prevents buildup and facilitates easy cleaning.
Designed to resist flex-fatigue and abrasions, the tubing is constructed with extra-heavy walls which can withstand a full vacuum at room temperature. It can be sterilized through conventional autoclave methods and ethylene oxide.
Applications include general laboratory, analytical instruments, and peristaltic and vacuum pumps. Ideal for condensers, desiccators, gas lines, and drain lines.
Call For Price
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Tubing ID (in) Tubing OD (in) Wall Thickness (in)
Availability Pricing
Tygon S3™ E-3603 Vacuum Tubing, 3/16" ID x 9/16" OD; 50 Ft 5011365 Mfr # ACF00015
COMPARE
3/16 9/16 3/16
Call For Price
Tygon S3™ E-3603 Vacuum Tubing, 1/4" ID x 5/8" OD; 50 Ft 5011366 Mfr # ACF00020
COMPARE
1/4 5/8 3/16
Call For Price
Tygon S3™ E-3603 Vacuum Tubing, 3/8" ID x 7/8" OD; 50 Ft 5011367 Mfr # ACF00030
COMPARE
3/8 7/8 1/4
Call For Price
Tygon S3™ E-3603 Vacuum Tubing, 1/2" ID x 1-1/8" OD; 50 Ft 5011368 Mfr # ACF00040
COMPARE
1/2 1 1/8 5/16
Call For Price
Tygon S3™ E-3603 Vacuum Tubing, 5/8" ID x 1-3/8" OD; 50 Ft 5011369 Mfr # ACF00049
COMPARE
5/8 1 3/8 3/8
Call For Price
Tygon S3™ E-3603 Vacuum Tubing, 3/4" ID x 1-1/2" OD; 50 Ft 5011370 Mfr # ACF00058
COMPARE
3/4 1 1/2 3/8
Call For Price
Tygon S3™ E-3603 Vacuum Tubing, 1" ID x 2" OD; 50 Ft 5011371 Mfr # ACF00066
COMPARE
1 2 1/2
Call For Price
Showing 1- 7 of 7